Anti-Neutrophils, blocking Rat Monoclonal Antibody [clone: Nimp-R14]

Lieferant: United States Biological
USBI045083EA 651 CHF
USBI045083
Anti-Neutrophils, blocking Rat Monoclonal Antibody [clone: Nimp-R14]
Antikörper
Type: Primary
Antigen: Neutrophils, blocking
Clonality: Monoclonal
Clone: Nimp-R14
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR