Anti-ACADSB Rabbit polyclonal antibody unconjugated

ABCAAB129710-100EA 830 CHF
ABCAAB129710-100
Anti-ACADSB Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: ACADSB (acyl-CoA dehydrogenase, short/branched chain)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR