Anti-GPR17 Rabbit polyclonal antibody unconjugated

ABCAAB111472-100EA 838 CHF
ABCAAB111472-100
Anti-GPR17 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: GPR17 (G protein-coupled receptor 17)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR