Anti-MYH4 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB111442-100EA 486.49 CHF
ABCAAB111442-100
Anti-MYH4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MYH4 (myosin, heavy chain 4, skeletal muscle)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR