Anti-CTAGE4 Rabbit polyclonal antibody unconjugated

ABCAAB111388-400EA 830 CHF
ABCAAB111388-400
Anti-CTAGE4 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: CTAGE4 (CTAGE family, member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR