Anti-MPC2 Rabbit polyclonal antibody unconjugated

ABCAAB111380-100EA 830 CHF
ABCAAB111380-100
Anti-MPC2 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: MPC2 (mitochondrial pyruvate carrier 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR