Anti-CT45A3 Rabbit polyclonal antibody unconjugated

ABCAAB111360-400EA 830 CHF
ABCAAB111360-400
Anti-CT45A3 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: CT45A3 (cancer/testis antigen family 45, member A3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR