Anti-CCNDBP1 Goat polyclonal antibody unconjugated

ABCAAB110868-50EA 830 CHF
ABCAAB110868-50
Anti-CCNDBP1 Goat polyclonal antibody unconjugated
Type: Primary
Antigen: CCNDBP1 (cyclin D-type binding-protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR