Anti-PREP Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB110857-50EA 534.19 CHF
ABCAAB110857-50
Anti-PREP Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PREP (prolyl endopeptidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR