Anti-TCEAL3/5/6 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB110856-100EA 870 CHF
ABCAAB110856-100
Anti-TCEAL3/5/6 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TCEAL3/5/6
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR