Anti-TRPC1 Goat Polyclonal Antibody
Lieferant: Abcam
ABCAAB110837-50EA 524.65 CHF
ABCAAB110837-50
Anti-TRPC1 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TRPC1 (transient receptor potential cation channel, subfamily C, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human