Anti-SPINK6 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB110830-100EA 505.57 CHF
ABCAAB110830-100
Anti-SPINK6 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SPINK6 (serine peptidase inhibitor, Kazal type 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR