Anti-CEP152 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB110825-100EA 524.65 CHF
ABCAAB110825-100
Anti-CEP152 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: CEP152 (centrosomal protein 152kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR