Anti-C1QL3 Rabbit polyclonal antibody unconjugated

ABCAAB107006-400EA 878 CHF
ABCAAB107006-400
Anti-C1QL3 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: C1QL3 (complement component 1, q subcomponent-like 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR