Anti-DAGLA Rabbit polyclonal antibody unconjugated

ABCAAB106979-400EA 900 CHF
ABCAAB106979-400
Anti-DAGLA Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: DAGLA (diacylglycerol lipase, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR