Anti-GPR17 Goat Polyclonal Antibody
Lieferant: Abcam
ABCAAB106781-100EA 524.65 CHF
ABCAAB106781-100
Anti-GPR17 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GPR17 (G protein-coupled receptor 17)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat