Anti-VCAM1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB106777-100EA 911 CHF
ABCAAB106777-100
Anti-VCAM1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: VCAM1 (vascular cell adhesion molecule 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR