Anti-ITGA5 Mouse Monoclonal Antibody (Peridinin Chlorophyll) [clone: CB49e]

Lieferant: Abcam
ABCAAB106776-50EA 290.94 CHF
ABCAAB106776-50
Anti-ITGA5 Mouse Monoclonal Antibody (Peridinin Chlorophyll) [clone: CB49e]
Antikörper
Type: Primary
Antigen: ITGA5 (integrin, alpha 5 (fibronectin receptor, alpha polypeptide))
Clonality: monoclonal
Clone: CB49e
Conjugation: Peridinin Chlorophyll
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR