Anti-CD79A Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: KT40]
Lieferant: Abcam
ABCAAB106765-100EA 524.65 CHF
ABCAAB106765-100
Anti-CD79A Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: KT40]
Antikörper
Type: Primary
Antigen: CD79A (CD79a molecule, immunoglobulin-associated alpha)
Clonality: monoclonal
Clone: KT40
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human