Anti-IgG/IgM Mouse Monoclonal Antibody [clone: KT100] (HRP (Horseradish Peroxidase))

Lieferant: Abcam
ABCAAB106762-100EA 496.03 CHF
ABCAAB106762-100
Anti-IgG/IgM Mouse Monoclonal Antibody [clone: KT100] (HRP (Horseradish Peroxidase))
Antikörper
Type: Secondary
Antigen: Rat IgG+IgM
Clonality: monoclonal
Clone: KT100
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR