Anti-SMC3 Rat Monoclonal Antibody [clone: KT75]

Lieferant: Abcam
ABCAAB106760-100EA 903 CHF
ABCAAB106760-100
Anti-SMC3 Rat Monoclonal Antibody [clone: KT75]
Antikörper
Type: Primary
Antigen: SMC3 (structural maintenance of chromosomes 3)
Clonality: monoclonal
Clone: KT75
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR