Anti-beta Sodium Potassium ATPase Goat polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB106744-100EA 911 CHF
ABCAAB106744-100
Anti-beta Sodium Potassium ATPase Goat polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: beta Sodium Potassium ATPase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR