Anti-MAEL Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB106713-100EA 534.19 CHF
ABCAAB106713-100
Anti-MAEL Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MAEL (maelstrom spermatogenic transposon silencer)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat