Anti-BST1 Mouse Monoclonal Antibody (APC (Allophycocyanin) [clone: 5Y/11B5]
Lieferant: Abcam
ABCAAB106712-50EA 252.79 CHF
ABCAAB106712-50
Anti-BST1 Mouse Monoclonal Antibody (APC (Allophycocyanin) [clone: 5Y/11B5]
Antikörper
Type: Primary
Antigen: BST1 (bone marrow stromal cell antigen 1)
Clonality: monoclonal
Clone: 5Y/11B5
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now