Anti-SCG2 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB104367-100EA 0 CHF
ABCAAB104367-100
Anti-SCG2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SCG2 (secretogranin II)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR