Anti-RBFOX3 Mouse Monoclonal Antibody [clone: 1B7]

Supplier: Abcam
ABCAAB104224-100EA 606.51 CHF
ABCAAB104224-100
Anti-RBFOX3 Mouse Monoclonal Antibody [clone: 1B7]
Antibodies
Type: Primary
Antigen: RBFOX3 (RNA binding protein, fox-1 homolog (C. elegans) 3)
Clonality: monoclonal
Clone: 1B7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR