Anti-SIGLEC7 Mouse Monoclonal Antibody [clone: MM0551-8D8]

Lieferant: Abcam
ABCAAB89483-100EA 903 CHF
ABCAAB89483-100
Anti-SIGLEC7 Mouse Monoclonal Antibody [clone: MM0551-8D8]
Antikörper
Type: Primary
Antigen: SIGLEC7 (sialic acid binding Ig-like lectin 7)
Clonality: monoclonal
Clone: MM0551-8D8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR