Anti-TNFRSF14 Mouse Monoclonal Antibody [clone: MM0332-5S1]

Lieferant: Abcam
ABCAAB89479-100EA 0 CHF
ABCAAB89479-100
Anti-TNFRSF14 Mouse Monoclonal Antibody [clone: MM0332-5S1]
Antikörper
Type: Primary
Antigen: TNFRSF14 (tumor necrosis factor receptor superfamily, member 14)
Clonality: monoclonal
Clone: MM0332-5S1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR