Anti-CCL17 Mouse monoclonal antibody unconjugated [clone: MM0141-6J30]

Lieferant: Abcam
ABCAAB89463-100EA 903 CHF
ABCAAB89463-100
Anti-CCL17 Mouse monoclonal antibody unconjugated [clone: MM0141-6J30]
Antikörper
Type: Primary
Antigen: CCL17 (chemokine (C-C motif) ligand 17)
Clonality: monoclonal
Clone: MM0141-6J30
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR