Anti-ADARB1 Mouse Polyclonal Antibody
Lieferant: Abcam
ABCAAB89436-50EA 534.19 CHF
ABCAAB89436-50
Anti-ADARB1 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ADARB1 (adenosine deaminase, RNA-specific, B1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human