Anti-IgG Goat Polyclonal Antibody (Gold, 1.4 nm (ultra-small))

Lieferant: Abcam
ABCAAB30825-500EA 677 CHF
ABCAAB30825-500
Anti-IgG Goat Polyclonal Antibody (Gold, 1.4 nm (ultra-small))
Antikörper
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: 1.4 nm Gold (ultra-small)
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR