Anti-EGLN3 Rabbit polyclonal antibody unconjugated
Lieferant: Abcam
ABCAAB30782-100EA 496.03 CHF
ABCAAB30782-100
Anti-EGLN3 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: EGLN3 (egl-9 family hypoxia-inducible factor 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat