Anti-Papillomavirus 16 (E7) Mouse Monoclonal Antibody [clone: 289-17013]

Lieferant: Abcam
ABCAAB30731-100EA 911 CHF
ABCAAB30731-100
Anti-Papillomavirus 16 (E7) Mouse Monoclonal Antibody [clone: 289-17013]
Antikörper
Type: Primary
Antigen: Papillomavirus 16 (E7)
Clonality: Monoclonal
Clone: 289-17013
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR