Anti-SREBF2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB30682-250EA 572.35 CHF
ABCAAB30682-250
Anti-SREBF2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SREBF2 (sterol regulatory element binding transcription factor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR