Anti-MLH1 Rabbit Polyclonal Antibody

ABCAAB9144-50EA 762 CHF
ABCAAB9144-50
Anti-MLH1 Rabbit Polyclonal Antibody
Type: Primary
Antigen: MLH1 (mutL homolog 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR