Anti-TLR2 Mouse Monoclonal Antibody Biotin [clone: TL2.3]

ABCAAB9103-25EA 737 CHF
ABCAAB9103-25
Anti-TLR2 Mouse Monoclonal Antibody Biotin [clone: TL2.3]
Type: Primary
Antigen: TLR2 (toll-like receptor 2)
Clonality: monoclonal
Clone: TL2.3
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR