Anti-HTLV I p19 Mouse Monoclonal Antibody [clone: 45/6.11.1.3]

Lieferant: Abcam
ABCAAB9079-100EA 903 CHF
ABCAAB9079-100
Anti-HTLV I p19 Mouse Monoclonal Antibody [clone: 45/6.11.1.3]
Antikörper
Type: Primary
Antigen: HTLV I p19
Clonality: monoclonal
Clone: 45/6.11.1.3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR