Anti-HIV1 Reverse Transcriptase Mouse Monoclonal Antibody [clone: 39/4.12.2]

Supplier: Abcam
ABCAAB9066-100EA 466.86 CHF
ABCAAB9066-100
Anti-HIV1 Reverse Transcriptase Mouse Monoclonal Antibody [clone: 39/4.12.2]
Antibodies
Type: Primary
Antigen: HIV1 Reverse Transcriptase
Clonality: monoclonal
Clone: 39/4.12.2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR