Anti-HIST3H3 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB9045-100EA 462.65 CHF
ABCAAB9045-100
Anti-HIST3H3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HIST3H3 (histone cluster 3, H3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR