Anti-LYZ Sheep Polyclonal Antibody

ABCAAB9011-1EA 672 CHF
ABCAAB9011-1
Anti-LYZ Sheep Polyclonal Antibody
Type: Primary
Antigen: LYZ (lysozyme)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR