Anti-ITGA3B/6B Mouse Monoclonal Antibody [clone: PB36]

ABCAAB8987-50EA 660 CHF
ABCAAB8987-50
Anti-ITGA3B/6B Mouse Monoclonal Antibody [clone: PB36]
Type: Primary
Antigen: ITGA3B/6B (integrin, alpha 3B/6B)
Clonality: monoclonal
Clone: PB36
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR