Anti-LMNB2 Mouse Monoclonal Antibody [clone: LN43]

Supplier: Abcam
ABCAAB8983-100EA 497.99 CHF
ABCAAB8983-100
Anti-LMNB2 Mouse Monoclonal Antibody [clone: LN43]
Antibodies
Type: Primary
Antigen: LMNB2 (lamin B2)
Clonality: monoclonal
Clone: LN43
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR