Anti-VIM Mouse Monoclonal Antibody [clone: RV203]

Supplier: Abcam
ABCAAB8979-100EA 456.49 CHF
ABCAAB8979-100
Anti-VIM Mouse Monoclonal Antibody [clone: RV203]
Antibodies
Type: Primary
Antigen: VIM (vimentin)
Clonality: monoclonal
Clone: RV203
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR