Anti-HP Sheep Polyclonal Antibody

Lieferant: Abcam
ABCAAB8968-1EA 911 CHF
ABCAAB8968-1
Anti-HP Sheep Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HP (haptoglobin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR