Anti-APOA1 Sheep Polyclonal Antibody

ABCAAB8945-1EA 0 CHF
ABCAAB8945-1
Anti-APOA1 Sheep Polyclonal Antibody
Type: Primary
Antigen: APOA1 (apolipoprotein A-I)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR