Anti-AKT1 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB8932-100EA 518.74 CHF
ABCAAB8932-100
Anti-AKT1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: AKT1 (v-akt murine thymoma viral oncogene homolog 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR