Anti-Dopamine Rabbit Polyclonal Antibody (Unconjugated)

Supplier: Abcam
ABCAAB8888-50EA 962 CHF
ABCAAB8888-50
Anti-Dopamine Rabbit Polyclonal Antibody (Unconjugated)
Antibodies
Type: Primary
Antigen: Dopamine
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR