Anti-NTRK1 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB8871-100EA 932 CHF
ABCAAB8871-100
Anti-NTRK1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NTRK1 (neurotrophic tyrosine kinase, receptor, type 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR