Anti-Factor I Sheep Polyclonal Antibody

Supplier: Abcam
ABCAAB8843-1EA 924 CHF
ABCAAB8843-1
Anti-Factor I Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Factor I
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR