Anti-KRT16 Mouse Monoclonal Antibody [clone: LL025]

ABCAAB8741-500EA 741 CHF
ABCAAB8741-500
Anti-KRT16 Mouse Monoclonal Antibody [clone: LL025]
Type: Primary
Antigen: KRT16 (keratin 16)
Clonality: monoclonal
Clone: LL025
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR